Product Details

SNP ID
rs61743475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20860321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTAGGGAACCGTGCACAGATTCC[A/G]TCAATGCTGTCTGCACTTATTGCTT
Phenotype
MIM: 616167
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCUN1D3 PubMed Links

Gene Details

Gene
DCUN1D3
Gene Name
defective in cullin neddylation 1 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173475.3 740 Silent Mutation GAC,GAT D160D NP_775746.1

View Full Product Details