Product Details
- SNP ID
-
rs10483261
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:21888371 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGAAAAGCCCTGAGTTGATAATGT[C/T]CATATACTCCAATGGTGACAAAGAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRAV12-2
PubMed Links
Gene Details
- Gene
- TRAV12-2
- Gene Name
- T cell receptor alpha variable 12-2
There are no transcripts associated with this gene.
View Full Product Details