Product Details

SNP ID
rs116923550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76828182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGGGCTTGGCACCTTGAGGAC[C/T]TGGGACTCAGAGGAAGGGAGCAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC74A PubMed Links

Gene Details

Gene
LRRC74A
Gene Name
leucine rich repeat containing 74A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194287.2 Intron NP_919263.2
XM_011536479.1 Intron XP_011534781.1
XM_011536483.1 Intron XP_011534785.1
XM_011536487.1 Intron XP_011534789.1
XM_017021032.1 Intron XP_016876521.1
XM_017021033.1 Intron XP_016876522.1
XM_017021034.1 Intron XP_016876523.1
XM_017021035.1 Intron XP_016876524.1
XM_017021036.1 Intron XP_016876525.1
XM_017021037.1 Intron XP_016876526.1
XM_017021038.1 Intron XP_016876527.1
XM_017021039.1 Intron XP_016876528.1
XM_017021040.1 Intron XP_016876529.1
XM_017021041.1 Intron XP_016876530.1

View Full Product Details