Product Details

SNP ID
rs61741976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91233118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGCCTCCCTTTGGTAGATTCC[A/C]GAAATGTTTGCTGATGCCAACTTCT
Phenotype
MIM: 601404
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C14orf159 PubMed Links

Gene Details

Gene
C14orf159
Gene Name
chromosome 14 open reading frame 159
There are no transcripts associated with this gene.

Gene
GPR68
Gene Name
G protein-coupled receptor 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177676.1 2229 UTR 3 NP_001171147.1
NM_003485.3 2229 UTR 3 NP_003476.3
XM_005268110.4 2229 UTR 3 XP_005268167.1
XM_005268111.3 2229 UTR 3 XP_005268168.1
XM_005268112.3 2229 UTR 3 XP_005268169.1
XM_006720262.3 2229 UTR 3 XP_006720325.1
XM_011537196.2 2229 UTR 3 XP_011535498.1
XM_011537197.2 2229 UTR 3 XP_011535499.1
XM_011537198.2 2229 UTR 3 XP_011535500.1
XM_011537199.2 2229 UTR 3 XP_011535501.1

View Full Product Details