Product Details

SNP ID
rs115648467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56644031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTGTGCTGAACTAGGACCCAGT[G/T]CTCTGTCCAGCATTGAGCTGCCTGA
Phenotype
MIM: 156350 MIM: 156349 MIM: 156356
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MT1A PubMed Links

Gene Details

Gene
MT1A
Gene Name
metallothionein 1A
There are no transcripts associated with this gene.

Gene
MT1B
Gene Name
metallothionein 1B
There are no transcripts associated with this gene.

Gene
MT1DP
Gene Name
metallothionein 1D, pseudogene
There are no transcripts associated with this gene.

Gene
MT1JP
Gene Name
metallothionein 1J, pseudogene
There are no transcripts associated with this gene.

View Full Product Details