Product Details

SNP ID
rs12460305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1278862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCAGTGCCCAGCAACCCCCTG[C/T]TCCACCGCTCATTCCCCTGCTGGCC
Phenotype
MIM: 602649 MIM: 602756
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf24 PubMed Links

Gene Details

Gene
C19orf24
Gene Name
chromosome 19 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017914.3 513 UTR 3 NP_060384.3
Gene
CIRBP
Gene Name
cold inducible RNA binding protein
There are no transcripts associated with this gene.

Gene
CIRBP-AS1
Gene Name
CIRBP antisense RNA 1
There are no transcripts associated with this gene.

Gene
EFNA2
Gene Name
ephrin A2
There are no transcripts associated with this gene.

View Full Product Details