Product Details

SNP ID
rs76673715
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:151286258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGACATCTCAGTTTCTCCTCCTC[C/G]TTCCTGTTTTCATCAGGTCTGGGAT
Phenotype
MIM: 602758 MIM: 610568
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100507670 PubMed Links

Gene Details

Gene
LOC100507670
Gene Name
uncharacterized LOC100507670
There are no transcripts associated with this gene.

Gene
PI4KB
Gene Name
phosphatidylinositol 4-kinase beta
There are no transcripts associated with this gene.

Gene
ZNF687
Gene Name
zinc finger protein 687
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304763.1 Intron NP_001291692.1
NM_001304764.1 Intron NP_001291693.1
NM_020832.2 Intron NP_065883.1
XM_005245366.3 Intron XP_005245423.1
XM_011509811.2 Intron XP_011508113.1
XM_011509812.2 Intron XP_011508114.1
XM_011509813.2 Intron XP_011508115.1

View Full Product Details