Product Details

SNP ID
rs56261530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4280210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCGAGGACCCCTATGAGGACGC[C/G]GAGAGCCGCTTGGAGCCGGACCCCG
Phenotype
MIM: 610481
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SHD PubMed Links

Gene Details

Gene
SHD
Gene Name
Src homology 2 domain containing transforming protein D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020209.3 1610 Silent Mutation GCC,GCG A49A NP_064594.3

View Full Product Details