Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022092.2 | 406 | Intron | NP_071375.1 | ||
XM_005255470.1 | 406 | Intron | XP_005255527.1 | ||
XM_005255471.3 | 406 | Silent Mutation | TCC,TCT | S115S | XP_005255528.1 |
XM_011522572.1 | 406 | Intron | XP_011520874.1 | ||
XM_011522573.1 | 406 | Intron | XP_011520875.1 | ||
XM_017023532.1 | 406 | Intron | XP_016879021.1 | ||
XM_017023533.1 | 406 | Intron | XP_016879022.1 | ||
XM_017023534.1 | 406 | Intron | XP_016879023.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001324086.1 | 406 | Intron | NP_001311015.1 | ||
NM_001324410.1 | 406 | Intron | NP_001311339.1 | ||
NM_001324411.1 | 406 | Intron | NP_001311340.1 | ||
NM_001324412.1 | 406 | Intron | NP_001311341.1 | ||
NM_001324413.1 | 406 | Intron | NP_001311342.1 | ||
NM_001324414.1 | 406 | Intron | NP_001311343.1 | ||
NM_001324415.1 | 406 | Intron | NP_001311344.1 | ||
NM_058192.2 | 406 | Intron | NP_478072.1 |