Product Details

SNP ID
rs8111802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9854147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATGAAAAGGGCCCCCAGCCCCC[A/G]GAGGCCCCCCAGGCAGCAGCCCGAG
Phenotype
MIM: 601052
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFM2 PubMed Links

Gene Details

Gene
OLFM2
Gene Name
olfactomedin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304347.1 1564 UTR 3 NP_001291276.1
NM_001304348.1 1564 UTR 3 NP_001291277.1
NM_058164.3 1564 UTR 3 NP_477512.1
Gene
PIN1
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1
There are no transcripts associated with this gene.

View Full Product Details