Product Details
- SNP ID
-
rs7485577
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:57222278 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCGCCAGGATCAGAGAAGCACAGAC[A/G]GCACTTTGTCTGAGATCAGAGATGG
- Phenotype
-
MIM: 107770
MIM: 604637
MIM: 138450
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LRP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149103307] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRP1
- Gene Name
- LDL receptor related protein 1
There are no transcripts associated with this gene.
- Gene
- NXPH4
- Gene Name
- neurexophilin 4
- Gene
- SHMT2
- Gene Name
- serine hydroxymethyltransferase 2
There are no transcripts associated with this gene.
View Full Product Details