Product Details

SNP ID
rs10022237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87307076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGTGGGTGAAAGATGGAATGAG[C/T]AAAGGGGCAGGGAAGATCTGGCTGA
Phenotype
MIM: 612127
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD17B13 PubMed Links

Gene Details

Gene
HSD17B13
Gene Name
hydroxysteroid 17-beta dehydrogenase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136230.2 Intron NP_001129702.1
NM_178135.4 Intron NP_835236.2
Gene
MIR5705
Gene Name
microRNA 5705
There are no transcripts associated with this gene.

View Full Product Details