Product Details

SNP ID
rs4442371
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:10056311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTACACAAAGGGGAAACTGAGC[C/T]TAAGAGGAAGATTAAAAGAGCAAAG
Phenotype
MIM: 613565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE4B PubMed Links

Gene Details

Gene
UBE4B
Gene Name
ubiquitination factor E4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105562.2 Intron NP_001099032.1
NM_006048.4 Intron NP_006039.2
XM_005263422.1 Intron XP_005263479.1
XM_011540488.2 Intron XP_011538790.1
XM_011540489.2 Intron XP_011538791.1
XM_011540490.1 Intron XP_011538792.1
XM_017000035.1 Intron XP_016855524.1
XM_017000036.1 Intron XP_016855525.1

View Full Product Details