Product Details

SNP ID
rs2943332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:25693648 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTGTGTTAAGGTCAGAGCAAAAT[C/G]AATAGGGAACAGTTACAAAGATCAT
Phenotype
MIM: 604059
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HS3ST4 PubMed Links

Gene Details

Gene
HS3ST4
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006040.2 Intron NP_006031.2

View Full Product Details