Product Details
- SNP ID
-
rs2175288
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:47327060 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTAACTCAGATGTGAGTTGAAAAT[A/G]TGAAAATTTGAGGAATTTCTAAGTG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EFCAB13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76421815,rs78024734] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EFCAB13
- Gene Name
- EF-hand calcium binding domain 13
- Gene
- THCAT158
- Gene Name
- thyroid cancer-associated transcript 158
There are no transcripts associated with this gene.
View Full Product Details