Product Details
- SNP ID
-
rs35624339
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:160763429 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAAACTGATATTTTTCTAGCTAG[C/T]TTAAAAAAATAAATAATTCACAACT
- Phenotype
-
MIM: 611931
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPM1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77829144] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPM1L
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1L
View Full Product Details