Product Details

SNP ID
rs13271442
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:70672934 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTTGTGTTTTCTCTTGTTGAGT[C/T]TGAGTTCCTTGTATGTTCTGGATAT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LACTB2 PubMed Links

Gene Details

Gene
LACTB2
Gene Name
lactamase beta 2
There are no transcripts associated with this gene.

Gene
LACTB2-AS1
Gene Name
LACTB2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
XKR9
Gene Name
XK related 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011720.1 Intron NP_001011720.1
NM_001287258.1 Intron NP_001274187.1
NM_001287259.1 Intron NP_001274188.1
NM_001287260.1 Intron NP_001274189.1
XM_006716447.3 Intron XP_006716510.1
XM_011517524.2 Intron XP_011515826.1
XM_011517525.2 Intron XP_011515827.1
XM_011517526.2 Intron XP_011515828.1
XM_011517527.2 Intron XP_011515829.1
XM_017013404.1 Intron XP_016868893.1
XM_017013405.1 Intron XP_016868894.1

View Full Product Details