Product Details

SNP ID
rs757917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:75651492 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTAGTGCAGTCATATACCATCTC[A/G]GTTACTCTTCGTCTGTAGATAGGTG
Phenotype
MIM: 611832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL19 PubMed Links

Gene Details

Gene
MRPL19
Gene Name
mitochondrial ribosomal protein L19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014763.3 Intron NP_055578.2

View Full Product Details