Product Details

SNP ID
rs1848069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:43375857 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGATGAATTTAATGCCATATAATA[C/T]TTTTTGTTTGTTCATTTTTGTTTGT
Phenotype
MIM: 613726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANO10 PubMed Links

Gene Details

Gene
ANO10
Gene Name
anoctamin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204831.1 Intron NP_001191760.1
NM_001204832.1 Intron NP_001191761.1
NM_001204833.1 Intron NP_001191762.1
NM_001204834.1 Intron NP_001191763.1
NM_018075.3 Intron NP_060545.3
XM_011533885.2 Intron XP_011532187.2
XM_011533887.2 Intron XP_011532189.1
XM_011533889.2 Intron XP_011532191.1
XM_011533890.2 Intron XP_011532192.1
XM_017006716.1 Intron XP_016862205.1
XM_017006717.1 Intron XP_016862206.1
XM_017006718.1 Intron XP_016862207.1
XM_017006719.1 Intron XP_016862208.1
XM_017006720.1 Intron XP_016862209.1
XM_017006721.1 Intron XP_016862210.1
XM_017006722.1 Intron XP_016862211.1

View Full Product Details