Product Details
- SNP ID
-
rs12625616
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:873608 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGTCTTTGTCCCTCTCTGCCTTTC[A/G]TGTCTCTGGCCCTCCCTTCCTCTGG
- Phenotype
-
MIM: 603705
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANGPT4
PubMed Links
Gene Details
- Gene
- ANGPT4
- Gene Name
- angiopoietin 4
View Full Product Details