Product Details
- SNP ID
-
rs10237467
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:5878797 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGATTATATACTTTATGGAATGTG[A/G]TTTTACCACAATAATTAAAAAGAAC
- Phenotype
-
MIM: 164795
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OCM
PubMed Links
Gene Details
- Gene
- OCM
- Gene Name
- oncomodulin
View Full Product Details