Product Details
- SNP ID
-
rs3801051
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:4997629 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCTGGCCACCTCTAAAATGATTGG[C/T]ATAGGGGCTTCCCCACCCCTTAGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF216P1
PubMed Links
Gene Details
- Gene
- RNF216P1
- Gene Name
- ring finger protein 216 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details