Product Details
- SNP ID
-
rs10802805
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
52
- Location
-
Chr.1:235549326 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACAGAAAAGCATCTCACAGGGGAA[A/G]GTAAGGAAGAGAGGAGGAAAAGGAA
- Phenotype
-
MIM: 604388
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GNG4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116218362] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNG4
- Gene Name
- G protein subunit gamma 4
View Full Product Details