Product Details
- SNP ID
-
rs2623064
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:103109635 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGTCATCCAAGTCTCTTGATACA[A/C]TCTCATCCTATCTATCAAATTCACT
- Phenotype
-
MIM: 117143
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BDH2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72665049] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BDH2
- Gene Name
- 3-hydroxybutyrate dehydrogenase, type 2
There are no transcripts associated with this gene.
- Gene
- CENPE
- Gene Name
- centromere protein E
View Full Product Details