Product Details

SNP ID
rs1074204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43716311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCACCGCAGACCATACTAGGGA[A/G]AGGGCACACCCAGGATAGGAGGCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRGC PubMed Links

Gene Details

Gene
IRGC
Gene Name
immunity related GTPase cinema
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019612.3 Intron NP_062558.1
XM_005259058.3 Intron XP_005259115.1
XM_006723283.3 Intron XP_006723346.1
XM_011527118.2 Intron XP_011525420.1

View Full Product Details