Product Details

SNP ID
rs4634313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180235722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAGTTTAAAGAAAACAAGATGT[A/C]CACATAAGTGTCTTGGTTTTAGCCG
Phenotype
MIM: 602896
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAPK9 PubMed Links

Gene Details

Gene
MAPK9
Gene Name
mitogen-activated protein kinase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135044.1 2266 Intron NP_001128516.1
NM_001308244.1 2266 Intron NP_001295173.1
NM_002752.4 2266 UTR 3 NP_002743.3
NM_139068.2 2266 UTR 3 NP_620707.1
NM_139069.2 2266 UTR 3 NP_620708.1
NM_139070.2 2266 UTR 3 NP_620709.1
XM_005265940.4 2266 UTR 3 XP_005265997.1
XM_006714891.2 2266 UTR 3 XP_006714954.1
XM_017009638.1 2266 UTR 3 XP_016865127.1
XM_017009639.1 2266 UTR 3 XP_016865128.1
XM_017009640.1 2266 UTR 3 XP_016865129.1
XM_017009641.1 2266 UTR 3 XP_016865130.1
XM_017009642.1 2266 Intron XP_016865131.1
XM_017009643.1 2266 Intron XP_016865132.1

View Full Product Details