Product Details

SNP ID
rs2637725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38667899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTTTAGAAAAATCATTGAAGCA[A/C]AGCCTAAGACATTGAGAAAATCCCG
Phenotype
MIM: 609392
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLF3 PubMed Links

Gene Details

Gene
KLF3
Gene Name
Kruppel like factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016531.5 Intron NP_057615.3
XM_017008279.1 Intron XP_016863768.1
Gene
KLF3-AS1
Gene Name
KLF3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details