Product Details

SNP ID
rs1042282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:846183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAACCTCAGTGCCTCCGTCGCC[A/T]CAGTCCAGCTGCCACAGCAGGACCA
Phenotype
MIM: 130130 MIM: 177020
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ELANE PubMed Links

Gene Details

Gene
ELANE
Gene Name
elastase, neutrophil expressed
There are no transcripts associated with this gene.

Gene
PRTN3
Gene Name
proteinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002777.3 450 Missense Mutation ACA,TCA T136S NP_002768.3
XM_011528136.1 450 Missense Mutation ACA,TCA T136S XP_011526438.1

View Full Product Details