Product Details

SNP ID
rs1131347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178206812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGCTATGAAGAAGGACCCGGT[C/G]AAGAAAATCTTCGTTGGGGGTCTGA
Phenotype
MIM: 602688 MIM: 614683
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPAB PubMed Links

Gene Details

Gene
HNRNPAB
Gene Name
heterogeneous nuclear ribonucleoprotein A/B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004499.3 716 Silent Mutation GTC,GTG V153V NP_004490.2
NM_031266.2 716 Silent Mutation GTC,GTG V153V NP_112556.2
Gene
PHYKPL
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278346.1 716 Intron NP_001265275.1
NM_153373.3 716 Intron NP_699204.1

View Full Product Details