Product Details

SNP ID
rs35643944
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:21920280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCGCCGCCTTCCTCTCACGCCC[C/G]GGAATACGAGAGACCCCCCAGCAAC
Phenotype
MIM: 604997 MIM: 606140
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DOK2 PubMed Links

Gene Details

Gene
DOK2
Gene Name
docking protein 2
There are no transcripts associated with this gene.

Gene
XPO7
Gene Name
exportin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015024.4 Intron NP_055839.3
XM_017013244.1 Intron XP_016868733.1
XM_017013245.1 Intron XP_016868734.1
XM_017013246.1 Intron XP_016868735.1
XM_017013247.1 Intron XP_016868736.1

View Full Product Details