Product Details

SNP ID
rs9930284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69344170 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAATAATGGAAAACTTCAGTACT[C/T]AAACTAGAGAGCTCTTCCAGAGGAC
Phenotype
MIM: 606979
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COG8 PubMed Links

Gene Details

Gene
COG8
Gene Name
component of oligomeric golgi complex 8
There are no transcripts associated with this gene.

Gene
NIP7
Gene Name
NIP7, nucleolar pre-rRNA processing protein
There are no transcripts associated with this gene.

Gene
TMED6
Gene Name
transmembrane p24 trafficking protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144676.3 Intron NP_653277.2

View Full Product Details