Product Details
- SNP ID
-
rs7555491
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
49
- Location
-
Chr.1:223707187 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAAAGAAAAATACTACTGAAAAAG[A/G]TTCCATTGCCATGGCCCTCTGAGAC
- Phenotype
-
MIM: 114230
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN2
PubMed Links
Gene Details
- Gene
- CAPN2
- Gene Name
- calpain 2
View Full Product Details