Product Details

SNP ID
rs2672603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122457318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTCCCGGGTGATAGGCATTA[A/C]CTAAAATTAAATAAAAATTCAGATC
Phenotype
MIM: 611313 MIM: 602194
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARMS2 PubMed Links

Gene Details

Gene
ARMS2
Gene Name
age-related maculopathy susceptibility 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099667.1 774 UTR 3 NP_001093137.1
Gene
HTRA1
Gene Name
HtrA serine peptidase 1
There are no transcripts associated with this gene.

View Full Product Details