Product Details

SNP ID
rs11214981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114409016 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGATTGCCTGCTGTTTAGAATAT[C/T]AGTTCTTTAATTTTGAGCATCCTGA
Phenotype
MIM: 612413
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf71 PubMed Links

Gene Details

Gene
C11orf71
Gene Name
chromosome 11 open reading frame 71
There are no transcripts associated with this gene.

Gene
RBM7
Gene Name
RNA binding motif protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286045.1 1905 UTR 3 NP_001272974.1
NM_001286046.1 1905 UTR 3 NP_001272975.1
NM_001286047.1 1905 UTR 3 NP_001272976.1
NM_001286048.1 1905 UTR 3 NP_001272977.1
NM_016090.3 1905 UTR 3 NP_057174.1

View Full Product Details