Product Details
- SNP ID
-
rs10803130
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
33
- Location
-
Chr.1:243258102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATACATTTTAAAATTAGTTTGAA[A/G]TAAAATTTAAATTTAGTGTGATAAC
- Phenotype
-
MIM: 613023
MIM: 613524
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP170
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116232091] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP170
- Gene Name
- centrosomal protein 170
There are no transcripts associated with this gene.
- Gene
- SDCCAG8
- Gene Name
- serologically defined colon cancer antigen 8
View Full Product Details