Product Details

SNP ID
rs7430698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100501747 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCCCTCAGTGCCGCAGTTATTT[C/T]GTGGTAGTAACATAGGTGTTTAGGA
Phenotype
MIM: 616928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM45A PubMed Links

Gene Details

Gene
TMEM45A
Gene Name
transmembrane protein 45A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018004.2 Intron NP_060474.1
XM_005247569.2 Intron XP_005247626.1
XM_017006715.1 Intron XP_016862204.1

View Full Product Details