Product Details

SNP ID
rs35602124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75004750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTTCTTACAGAAGGGACAATGGA[A/G]AACATTGCAGCCCAGGCTCTGGAGC
Phenotype
MIM: 606454 MIM: 604395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2B2 PubMed Links

Gene Details

Gene
EIF2B2
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014239.3 529 Silent Mutation GAA,GAG E149E NP_055054.1
Gene
MLH3
Gene Name
mutL homolog 3
There are no transcripts associated with this gene.

View Full Product Details