Product Details

SNP ID
rs4580644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15783578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTAACACATTGCTTATCACCTAA[C/T]AGTCTCTTAATAGAAGCTTGCTATA
Phenotype
MIM: 600387 MIM: 107270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BST1 PubMed Links

Gene Details

Gene
BST1
Gene Name
bone marrow stromal cell antigen 1
There are no transcripts associated with this gene.

Gene
CD38
Gene Name
CD38 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001775.3 Intron NP_001766.2

View Full Product Details