Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001029851.2 | Intron | NP_001025022.1 | |||
NM_001029852.2 | Intron | NP_001025023.1 | |||
NM_001029853.2 | Intron | NP_001025024.1 | |||
NM_001029854.2 | Intron | NP_001025025.1 | |||
NM_003719.3 | Intron | NP_003710.1 | |||
XM_005248621.4 | Intron | XP_005248678.1 | |||
XM_005248623.4 | Intron | XP_005248680.1 | |||
XM_005248624.4 | Intron | XP_005248681.1 | |||
XM_006714725.3 | Intron | XP_006714788.1 | |||
XM_006714726.3 | Intron | XP_006714789.1 | |||
XM_011543699.2 | Intron | XP_011542001.1 | |||
XM_011543700.2 | Intron | XP_011542002.1 | |||
XM_011543701.2 | Intron | XP_011542003.1 | |||
XM_011543702.2 | Intron | XP_011542004.1 | |||
XM_011543703.2 | Intron | XP_011542005.1 | |||
XM_011543704.2 | Intron | XP_011542006.1 | |||
XM_017010003.1 | Intron | XP_016865492.1 | |||
XM_017010004.1 | Intron | XP_016865493.1 | |||
XM_017010005.1 | Intron | XP_016865494.1 | |||
XM_017010006.1 | Intron | XP_016865495.1 | |||
XM_017010007.1 | Intron | XP_016865496.1 | |||
XM_017010008.1 | Intron | XP_016865497.1 |