Product Details

SNP ID
rs2683451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169528022 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTGTTATCACTTGCTTGAGAAAT[A/G]GAACACATTTCATGGAAATTAAACA
Phenotype
MIM: 607701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FASTKD1 PubMed Links

Gene Details

Gene
FASTKD1
Gene Name
FAST kinase domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281476.2 4753 Intron NP_001268405.1
NM_001322046.1 4753 Intron NP_001308975.1
NM_001322048.1 4753 Intron NP_001308977.1
NM_001322049.1 4753 Intron NP_001308978.1
NM_024622.5 4753 Intron NP_078898.3
XM_006712750.3 4753 UTR 3 XP_006712813.1
XM_006712751.3 4753 UTR 3 XP_006712814.1
XM_017004911.1 4753 UTR 3 XP_016860400.1
XM_017004912.1 4753 UTR 3 XP_016860401.1
Gene
KLHL41
Gene Name
kelch like family member 41
There are no transcripts associated with this gene.

View Full Product Details