Product Details

SNP ID
rs2573247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232480528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGGGAGATGAAGCCAGGCATC[C/T]GCCCCCTTGCCCCCCACCCAGCACA
Phenotype
MIM: 605896
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECEL1 PubMed Links

Gene Details

Gene
ECEL1
Gene Name
endothelin converting enzyme like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290787.1 Intron NP_001277716.1
NM_004826.3 Intron NP_004817.2

View Full Product Details