Product Details

SNP ID
rs2875812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237798842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCGTGGAGGCGGCAGCGGCGGC[C/G]GCTTTCTAGCGGATCCGAGGTGCGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RBM44 PubMed Links

Gene Details

Gene
RBM44
Gene Name
RNA binding motif protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080504.2 454 UTR 5 NP_001073973.2
XM_005246075.1 454 UTR 5 XP_005246132.1
XM_011511152.2 454 UTR 5 XP_011509454.1
XM_011511153.2 454 UTR 5 XP_011509455.1
XM_011511154.2 454 UTR 5 XP_011509456.1
XM_011511155.1 454 Intron XP_011509457.1
XM_017004054.1 454 UTR 5 XP_016859543.1
XM_017004055.1 454 UTR 5 XP_016859544.1
XM_017004056.1 454 Intron XP_016859545.1
XM_017004057.1 454 UTR 5 XP_016859546.1
XM_017004058.1 454 Intron XP_016859547.1

View Full Product Details