Product Details

SNP ID
rs12795672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8225007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCAGGTCTCCTGTGTTGGTGAG[G/T]GTAGGGTTAGGGGGGTTGGTGGAGA
Phenotype
MIM: 186921
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LMO1 PubMed Links

Gene Details

Gene
LMO1
Gene Name
LIM domain only 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270428.1 Intron NP_001257357.1
NM_002315.2 Intron NP_002306.1
XM_006718228.3 Intron XP_006718291.1
XM_011520098.2 Intron XP_011518400.1
XM_011520099.2 Intron XP_011518401.1
XM_011520100.1 Intron XP_011518402.1

View Full Product Details