Product Details

SNP ID
rs12127009
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:185061415 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGAAAGCTTCTGATCTCAGGCA[A/G]AAACCAACCCACCAAACAAAAAAAA
Phenotype
MIM: 608985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF2 PubMed Links

Gene Details

Gene
RNF2
Gene Name
ring finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007212.3 Intron NP_009143.1
XM_005245413.3 Intron XP_005245470.1
XM_011509851.2 Intron XP_011508153.1
XM_011509852.2 Intron XP_011508154.1

View Full Product Details