Product Details
- SNP ID
-
rs2516708
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:30260944 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCTCCCTCACACTCAGTGTGTCC[A/G]TGGCTCCGATTCCAGCTCTTCTGAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HCG17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113639903] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HCG17
- Gene Name
- HLA complex group 17 (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- HLA-L
- Gene Name
- major histocompatibility complex, class I, L (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details