Product Details

SNP ID
rs13096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25206907 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTTAATGTGTACAGTAATTGTCC[C/T]AAAAGAATCACAGTTATGCCAAATA
Phenotype
MIM: 190070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRAS PubMed Links

Gene Details

Gene
KRAS
Gene Name
KRAS proto-oncogene, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004985.4 4259 UTR 3 NP_004976.2
NM_033360.3 4259 UTR 3 NP_203524.1
XM_006719069.3 4259 Intron XP_006719132.1
XM_011520653.2 4259 Intron XP_011518955.1
XM_017019292.1 4259 Intron XP_016874781.1
XM_017019293.1 4259 UTR 3 XP_016874782.1
Gene
LYRM5
Gene Name
LYR motif containing 5
There are no transcripts associated with this gene.

View Full Product Details