Product Details

SNP ID
rs35492502
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:217632882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCATGATGTTCACATCCTTGTAT[A/G]GAATATAGGCTTTTAAAAAATAATT
Phenotype
MIM: 616836 MIM: 611032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH2 PubMed Links

Gene Details

Gene
GPATCH2
Gene Name
G-patch domain containing 2
There are no transcripts associated with this gene.

Gene
SPATA17
Gene Name
spermatogenesis associated 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138796.3 Intron NP_620151.1
XM_005273052.1 Intron XP_005273109.1
XM_006711165.3 Intron XP_006711228.1
XM_011509194.2 Intron XP_011507496.1

View Full Product Details