Product Details

SNP ID
rs2420532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14538452 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGAGAGGGACCCACGTTAGGCAG[A/G]TAAGGACAGATCAGCATCCCCAGGG
Phenotype
MIM: 604572 MIM: 610057
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB1 PubMed Links

Gene Details

Gene
DNAJB1
Gene Name
DnaJ heat shock protein family (Hsp40) member B1
There are no transcripts associated with this gene.

Gene
MIR639
Gene Name
microRNA 639
There are no transcripts associated with this gene.

Gene
TECR
Gene Name
trans-2,3-enoyl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321170.1 Intron NP_001308099.1
NM_138501.5 Intron NP_612510.1
XM_006722945.1 Intron XP_006723008.1
XM_011528442.2 Intron XP_011526744.1

View Full Product Details