Product Details

SNP ID
rs3813611
Assay Type
Functionally Tested
NCBI dbSNP Submissions
41
Location
Chr.1:161098450 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CCGGTAGGGGGAGGTTCCCGGGGAA[A/G]CCCGCGGAAGGCGAGGTGCCTGGCC
Phenotype
MIM: 609607 MIM: 613466
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KLHDC9 PubMed Links

Gene Details

Gene
KLHDC9
Gene Name
kelch domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007255.2 60 UTR 5 NP_001007256.1
NM_152366.4 60 UTR 5 NP_689579.3
Gene
NECTIN4
Gene Name
nectin cell adhesion molecule 4
There are no transcripts associated with this gene.

Gene
PFDN2
Gene Name
prefoldin subunit 2
There are no transcripts associated with this gene.

View Full Product Details