Product Details

SNP ID
rs34413912
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:159856858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAAGAAAGGACCTCTCTCCCAG[C/T]AGCAAGACAGGGACACCATTAATTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf204 PubMed Links

Gene Details

Gene
C1orf204
Gene Name
chromosome 1 open reading frame 204
There are no transcripts associated with this gene.

Gene
VSIG8
Gene Name
V-set and immunoglobulin domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013661.1 Intron NP_001013683.1

View Full Product Details